Modular Basket System
Modular basket system type 80, chamfer 20×20 mm, dimension: 455×105×55 mm
Catalog # ELMA1111838
Supplier: ELMA SCHMIDBAUER
Jump to:
Product Details & Documents
Modular basket system for Elmasonic P, Elmasonic Select and EASY series ultrasonic baths.
- Stainless steel with plastic handles (10 to 120 and 180 to 300) or Rilsan coating (150)
- Ergonomic handles
- Autoclavable
- Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
- Square openings in the side walls for mounting silicone holders
Hole diameter bottom (mm): 11×15
Hole diameter sides (mm): 10×10
Specifications
- Description:Modular basket system type 80, chamfer 20×20 mm, dimension: 455×105×55 mm
- Seq:06
- Cat. No.:ELMA1111838
- Model:Type 80
- W×D×H:455×105×55 mm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Cover for round basket, stainless steel, Ext.Ø×H: 64×2 mmCover for round basket, stainless steel, Ext.Ø×H: 64×2 mmELMA3000006018Log in for availability1 * 1 ST - 14,00 €
Product Information
- Catalog #AvailabilityPrice
- Round basket with divisions creating 4 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmRound basket with divisions creating 4 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmELMA3000006013Log in for availability1 * 1 ST - 36,00 €
Product Information
- Catalog #AvailabilityPrice
- Round basket without divisions, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmRound basket without divisions, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmELMA3000006012Log in for availability1 * 1 ST - 22,00 €
Product Information
- Catalog #AvailabilityPrice
- Cover for minature round basket, stainless steel, Ext.Ø×H: 20×4 mmCover for minature round basket, stainless steel, Ext.Ø×H: 20×4 mmELMA3000006016Log in for availability1 * 1 ST - 18,00 €
Product Information
- Catalog #AvailabilityPrice
- Round basket with divisions creating 5 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmRound basket with divisions creating 5 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmELMA3000006014Log in for availability1 * 1 ST - 41,00 €
Product Information
- Catalog #AvailabilityPrice
- Minature round basket without divisions, stainless steel, Ext.Ø×H: 21×12 mm, Int.Ø×H: 18×11 mmMinature round basket without divisions, stainless steel, Ext.Ø×H: 21×12 mm, Int.Ø×H: 18×11 mmELMA3000006017Log in for availability1 * 1 ST - 14,00 €
Product Information
- Catalog #AvailabilityPrice
- Round basket without divisions, stainless steel, ¹/₁ height, Ext.Ø×H: 64×42 mm, Int.Ø×H: 60×41 mmRound basket without divisions, stainless steel, ¹/₁ height, Ext.Ø×H: 64×42 mm, Int.Ø×H: 60×41 mmELMA3000009104Log in for availability1 * 1 ST - 41,00 €
Product Information
- Catalog #AvailabilityPrice
- Round basket with divisions creating 9 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmRound basket with divisions creating 9 areas, stainless steel, ⅓ height, Ext.Ø×H: 64×16 mm, Int.Ø×H: 60×15 mmELMA3000006015Log in for availability1 * 1 ST - 64,00 €
Product Information
- Catalog #AvailabilityPrice
- Level set type L, hole Ø 11×15 mm, stainless steel, 250×195×1 mmLevel set type L, hole Ø 11×15 mm, stainless steel, 250×195×1 mmELMA1125227Log in for availability1 * 1 ST - 33,00 €
Product Information
- Catalog #AvailabilityPrice
- Level set type XL, hole Ø 11×15 mm, stainless steel, 285×250×1 mmLevel set type XL, hole Ø 11×15 mm, stainless steel, 285×250×1 mmELMA1125228Log in for availability1 * 1 ST - 36,00 €
Product Information
- Catalog #AvailabilityPrice
- Elma Pin 10×0772, plastic, Ext.Ø×H: 25×55 mm, Int.Ø×H: 5×40 mmElma Pin 10×0772, plastic, Ext.Ø×H: 25×55 mm, Int.Ø×H: 5×40 mmELMA1125213Log in for availability1 * 1 ST - 16,00 €
Product Information
- Catalog #AvailabilityPrice
- Elma Pin 10×0509, black, plastic, Ext.Ø×H: 30×35 mm, Int.Ø×H: 20×20 mmElma Pin 10×0509, black, plastic, Ext.Ø×H: 30×35 mm, Int.Ø×H: 20×20 mmELMA1125209Log in for availability1 * 1 ST - 16,00 €
Product Information
- Catalog #AvailabilityPrice
- Elma Pin 10×0158, plastic, Ext.Ø×H: 50×40 mm, Int.Ø×H: 40×25 mmElma Pin 10×0158, plastic, Ext.Ø×H: 50×40 mm, Int.Ø×H: 40×25 mmELMA1125210Log in for availability1 * 1 ST - 16,00 €
Product Information
- Catalog #AvailabilityPrice
- Elma Pin 10×0021R, plastic, Ext.Ø×H: 25×55 mm, Int.Ø×H: 15×40 mmElma Pin 10×0021R, plastic, Ext.Ø×H: 25×55 mm, Int.Ø×H: 15×40 mmELMA1125216Log in for availability1 * 1 ST - 16,00 €
Product Information
- Catalog #AvailabilityPrice
- Dividing wall set S high. 2 sets for basket sizes: 40/60/100/120/180 and 4 sets for basket sizes: 80/300, hole Ø 10×10 mm, stainless steel, 105×90×1 mmDividing wall set S high. 2 sets for basket sizes: 40/60/100/120/180 and 4 sets for basket sizes: 80/300, hole Ø 10×10 mm, stainless steel, 105×90×1 mmELMA1125221Log in for availability1 * 1 ST - 41,00 €
Product Information
- Catalog #AvailabilityPrice
- Elma starter set (contains: 3× each Elma pin 0741 and 0772 per set, 2× each Elma Pin 0509, 0158, 0767, 0613 and 0614, 0021R and 0001R per set, 2× silicone holder side comb and bottom comb each per set)Elma starter set (contains: 3× each Elma pin 0741 and 0772 per set, 2× each Elma Pin 0509, 0158, 0767, 0613 and 0614, 0021R and 0001R per set, 2× silicone holder side comb and bottom comb each per set)ELMA1124042Log in for availability1 * 1 ST - 51,00 €
Product Information
- Catalog #AvailabilityPrice
- Silicone holder set (10), Ext.Ø×H: 20×20 mm
Product Information
- Catalog #AvailabilityPrice
- Holder set, stainless steel
Product Information
- Catalog #AvailabilityPrice
- Level set type M, hole Ø 11×15 mm, stainless steel, 255×105×1 mmLevel set type M, hole Ø 11×15 mm, stainless steel, 255×105×1 mmELMA1125226Log in for availability1 * 1 ST - 16,00 €
Product Information
- Catalog #AvailabilityPrice
- Elma Pin 10×0741, plastic, Ext.Ø×H: 25×30 mm, Int.Ø×H: 4×15 mmElma Pin 10×0741, plastic, Ext.Ø×H: 25×30 mm, Int.Ø×H: 4×15 mmELMA1125214Log in for availability1 * 1 ST - 16,00 €
Product Information
- Catalog #AvailabilityPrice
- Elma Pin 10×0001R, plastic, Ext.Ø×H: 40×75 mm, Int.Ø×H: 30×65 mmElma Pin 10×0001R, plastic, Ext.Ø×H: 40×75 mm, Int.Ø×H: 30×65 mmELMA1125215Log in for availability1 * 1 ST - 16,00 €
Product Information
- Catalog #AvailabilityPrice
- Dividing wall set M. 2 sets for basket sizes: 100/120, hole Ø 10×10 mm, stainless steel, 200×60×1 mmDividing wall set M. 2 sets for basket sizes: 100/120, hole Ø 10×10 mm, stainless steel, 200×60×1 mmELMA1125223Log in for availability1 * 1 ST - 36,00 €
Product Information
- Catalog #AvailabilityPrice
- Silicone holder 10×bottom comb set, 105×10×35 mmSilicone holder 10×bottom comb set, 105×10×35 mmELMA1125217Log in for availability1 * 1 ST - 62,00 €
Product Information
- Catalog #AvailabilityPrice
- Dividing wall set S low. 1 set for basket size: 20; 2 sets for basket sizes: 30/40/60/100/120/180 and 4 sets for basket sizes: 80/150/300, hole Ø 10×10 mm, stainless steel, 105×50×1 mmDividing wall set S low. 1 set for basket size: 20; 2 sets for basket sizes: 30/40/60/100/120/180 and 4 sets for basket sizes: 80/150/300, hole Ø 10×10 mm, stainless steel, 105×50×1 mmELMA1125220Log in for availability1 * 1 ST - 26,00 €
Product Information
- Catalog #AvailabilityPrice
- Dividing wall set L low. 4 sets for basket sizes: 150, hole Ø 10×10 mm, stainless steel, 200×90×1 mmDividing wall set L low. 4 sets for basket sizes: 150, hole Ø 10×10 mm, stainless steel, 200×90×1 mmELMA1125222Log in for availability1 * 1 ST - 36,00 €
Product Information
- Catalog #AvailabilityPrice
- Dividing wall set L high. 2 sets for basket sizes: 180/300, hole Ø 10×10 mm, stainless steel, 255×130×1 mmDividing wall set L high. 2 sets for basket sizes: 180/300, hole Ø 10×10 mm, stainless steel, 255×130×1 mmELMA1125224Log in for availability1 * 1 ST - 41,00 €
Product Information
- Catalog #AvailabilityPrice
- Silicone holder 10×side comb set, 150×10×45 mmSilicone holder 10×side comb set, 150×10×45 mmELMA1125218Log in for availability1 * 1 ST - 62,00 €
Product Information
- Catalog #AvailabilityPrice
- Elma Pin 10×0613 and 0614, plastic, Ext.Ø×H: 55×45 mm and 75×45 mm, Int.Ø×H: 0,35×25 mm and 25×25 mmElma Pin 10×0613 and 0614, plastic, Ext.Ø×H: 55×45 mm and 75×45 mm, Int.Ø×H: 0,35×25 mm and 25×25 mmELMA1125212Log in for availability1 * 1 ST - 51,00 €
Product Information
- Catalog #AvailabilityPrice
- Elma Pin 10×0767, plastic, Ext.Ø×H: 85×65 mm, Int.Ø×H: 50×35 mmElma Pin 10×0767, plastic, Ext.Ø×H: 85×65 mm, Int.Ø×H: 50×35 mmELMA1125211Log in for availability1 * 1 ST - 16,00 €
Product Information
- Catalog #AvailabilityPrice
- Level set type S with one basket load, hole Ø 11×15 mm, stainless steel, 195×105×1 mmLevel set type S with one basket load, hole Ø 11×15 mm, stainless steel, 195×105×1 mmELMA1125225Log in for availability1 * 1 ST - 18,00 €