Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Acetone ≥99%, TECHNICAL

950 L
Catalog # 20063.930
Flammable
Irritant
Supplier:   VWR Chemicals
Acetone ≥99%, TECHNICAL
Catalog # 20063.930
Supplier:  VWR Chemicals
CAS Number:  67-64-1
Acetone ≥99%, TECHNICAL
Acetone ≥99%, TECHNICAL

950 L

CAS Number:  67-64-1
Price
Inquire For Price
Log in for availability
1 * 950 L
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • IR Spectrum:
    Passes test

Specifications

  • Pk:
    950 L

Specifications