D(-)-Lactic acid
Bottle, 500 g
Catalog # APOSOR960721-500G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: (R)-2-Hydroxypropanoic acid, D-Lactic Acid, (R)-(-)-2-Hydroxypropionic acid, (R)-(-)-Lactic acid, (R)-(-)-2-Hydroxypropanoic acid
- CAS Number:10326-41-7
- Formula:C₃H₆O₃
- MDL Number:MFCD00068311
- Molecular Weight:90.08 g/mol
- Storage Temperature:Freezer
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



