D-(+)-Biotin 99%
Bottle, 10 g
Catalog # APOSBIB0603-10G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
- CAS Number:58-85-5
- Formula:C₁₀H₁₆N₂O₃S
- MDL Number:MFCD00005541
- Molecular Weight:244.31 g/mol
- Melting Pt:231…233 °C
- Storage Temperature:Refrigerator
- EINECS:200-399-3
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Bottle
- Pk:10 g



