Order Entry
Germany
Orders LinkContactUsLinkComponent
2-(4-Bromophenyl)ethanol

2-(4-Bromophenyl)ethanol

Bottle, 100 g
Catalog # APOSOR30084-100G
Irritant
Supplier:   Apollo Scientific
2-(4-Bromophenyl)ethanol
Catalog # APOSOR30084-100G
Supplier:  Apollo Scientific
CAS Number:  4654-39-1
2-(4-Bromophenyl)ethanol

Bottle, 100 g

CAS Number:  4654-39-1
Price
210,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  4-Bromophenethyl alcohol
  • CAS Number:4654-39-1
  • Formula:C₈H₉BrO
  • MDL Number:MFCD00002897
  • Molecular Weight:201.06 g/mol
  • Boiling Pt:138 °C/9mmHg
  • Density:1.436 g/cm³ (25 °C)
  • Flash Pt:110 °C
  • Storage Temperature:Ambient
  • EINECS:225-093-7
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.