Order Entry
Germany
Orders LinkContactUsLinkComponent
5'-Bromo-2'-hydroxypropiophenone

5'-Bromo-2'-hydroxypropiophenone

Bottle, 1 g
Catalog # APOSOR22402-1G
Toxic
Supplier:   Apollo Scientific
5'-Bromo-2'-hydroxypropiophenone
Catalog # APOSOR22402-1G
Supplier:  Apollo Scientific
CAS Number:  17764-93-1
5'-Bromo-2'-hydroxypropiophenone

Bottle, 1 g

CAS Number:  17764-93-1
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  1-(5-Bromo-2-hydroxyphenyl)propan-1-one
  • CAS Number:17764-93-1
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD00218577
  • Molecular Weight:229.0706 g/mol
  • Melting Pt:76…80 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.