phi29 Polymerase
Catalog # APOSBIBL0054-1000U
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Storage at −20 °C. DNA Polymerase 10 U/μl. Highly processive recombinant polymerase with exceptional strand displacement activity, which allows for highly efficient isothermal DNA amplification. The enzyme is capable of up to 70 thousands base insertions per binding event. The polymerase also possesses a 3'→5' proofreading exonuclease activity acting preferentially on ssDNA or ssRNA. Extremely high yields of amplified DNA can be obtained even from minute amounts of a template.
Specifications
Specifications
- Cat. No.:APOSBIBL0054-1000U
- Enzyme name:phi29 Polymerase