SS-C-T-MERCK1, Sapphire tubes, OD1.5875mm(no more than 1/16)*ID0.8mm * L70mm. One end cut off at an angle, see DRW# IMG_20200304_162 443.OD polished, ID as grown 1 * 1 ST
Catalog # MSPPSS-C-T-MERCK1
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together