Order Entry
Germany
Orders LinkContactUsLinkComponent
GC septa

GC septa

BTO premium high-temperature GC septa, 6,4 mm, 400 °C
Catalog # SGEA298755
GC septa
Catalog # SGEA298755
GC septa
GC septa

BTO premium high-temperature GC septa, 6,4 mm, 400 °C

Price
107,20 €
Log in for availability
1 * 50 ST
Product Details & Documents
Specifications
  • Ø:
    6,4 mm
  • Max. temp.:
    400 °C
  • Description:
    BTO premium high-temperature GC septa
  • Seq:
    03