Order Entry
Germany
Orders LinkContactUsLinkComponent
GC septa

GC septa

BTO premium high-temperature centre guide GC septa, 12,7 mm, 400 °C
Catalog # SGEA298727
GC septa
Catalog # SGEA298727
GC septa
GC septa

BTO premium high-temperature centre guide GC septa, 12,7 mm, 400 °C

Price
107,20 €
Log in for availability
1 * 50 ST
Product Details & Documents
Specifications
  • Ø:
    12,7 mm
  • Max. temp.:
    400 °C
  • Description:
    BTO premium high-temperature centre guide GC septa
  • Seq:
    16
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.