N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanaminium chloride,90+% 1 * 25 g
Catalog # AMBDA1003341-25G
Supplier: AMBEED
N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanaminium chloride,90+% 1 * 25 g
Catalog # AMBDA1003341-25G
Supplier: AMBEED
Frequently Bought Together
(±)-2-Butanol, EMSURE® for analysis, Supelco®
Supplier: Merck
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
Diethyl ether ≥99% stabilised, GPR RECTAPUR® peroxide-free
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, SupraSolv® for GC-MS, Supelco®
Supplier: Merck
n-Heptane
Supplier: Merck
n-Hexane ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals