MELTING AGENT LiT/LiM/LiBr 50/50/1.0 High purity (99.98+%) pre-melted flux. flux. Composition: 49.5 % Lithium tetraborate, 49.5 % Li thium metaborate and 1.0 % lithium bromide. 1 * 1 kg
Catalog # MSPPC-0620-63
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together