ContactUsLinkComponent
Point Tester Method(s): ASTM D3278 ASTM D3828 ASTM D7236 ASTM D8174 ASTM E502 IP 523 IP 534 ISO 3679 ISO 3680 (Obs) EPA 1020 B CLP Regulations EC No 1272/2008 BS EN ISO 3679 & BS 2000-523 ASTM D1655 * -30C to 135C (no external cooler re 1 * 1 items
Catalog # STAN82100-2
Supplier:  STANHOPE SETA
Point Tester Method(s): ASTM D3278 ASTM D3828 ASTM D7236 ASTM D8174 ASTM E502 IP 523 IP 534 ISO 3679 ISO 3680 (Obs) EPA 1020 B CLP Regulations EC No 1272/2008 BS EN ISO 3679 & BS 2000-523 ASTM D1655 * -30C to 135C (no external cooler re 1 * 1 items
Catalog # STAN82100-2
Supplier:  STANHOPE SETA