Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SOLVENT BOTTLES 180ML GLASS 2/PKG 1 * 1 items
Catalog # GILI2954663
Supplier:
GILSON LCMS
Expand image
SOLVENT BOTTLES 180ML GLASS 2/PKG 1 * 1 items
Catalog # GILI2954663
Supplier:
GILSON LCMS