Order Entry
Germany
Orders LinkContactUsLinkComponent
Human Recombinant VEGF-121 | Avantor

Human Recombinant VEGF-121

100 µG
Catalog # STMC78127.1
Marketsource
Human Recombinant VEGF-121
Catalog # STMC78127.1
Marketsource
CAS Number:  
Human Recombinant VEGF-121
Human Recombinant VEGF-121

100 µG

Wechseln zu:
CAS Number:  
Price
1 090,00 €
Anmelden für die Verfügbarkeit
1 * 100 µG
Product Details & Documents

Vascular endothelial growth factor (VEGF-121) is a naturally-occurring VEGF-A splice variant involved in embryonic vasculogenesis and angiogenesis. VEGF binds to VEGFR-1 and VEGFR-2, and activates Raf/MEK/ERK and PI3K/AKT pathways. VEGF-121 is released as a freely diffusible protein by a variety of normal and transformed cells. It plays an important role in neurogenesis both in vitro and in vivo (Storkebaum et al.). It has neurotrophic effects on neurons of the central nervous system and promotes growth and survival of dopaminergic neurons and astrocytes. VEGF also promotes growth and survival of vascular endothelial cells, monocyte chemotaxis, and colony formation by granulocyte-macrophage progenitor cells (Ferrara et al.).

Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Spezifikationen:

Spezifikationen:

  • Pk:
    100 µG
  • Cat. No.:
    STMC78127.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    VEGF-121
  • Purity:
    ≥95%
  • Species:
    Human

Spezifikationen:

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen.