Order Entry
Germany
Orders LinkContactUsLinkComponent
Polysorbate 80 solution 0,1

Polysorbate 80 solution 0,1

Catalog # ABCAAB291111-100
Marketsource
Supplier:   Abcam
Polysorbate 80 solution 0,1
Catalog # ABCAAB291111-100
Supplier:  Abcam
Marketsource
CAS Number:  9005-65-6
Polysorbate 80 solution 0,1
CAS Number:  9005-65-6
Price
154,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Tween 80 Detergent, 10% Solution, ab291111 (previously known as Tween® 80, MegaPure™ Detergent, 10% Solution, 2126) is a non-ionic detergent useful for the extraction of membrane-bound proteins as well as isolation of nuclei from ma mMalian cell lines.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
  • CAS Number:9005-65-6
Specifications

Specification Test Results

  • Constituents::
    10% Tween 80