This In Vitro MicroFlow kit allows users to analyze in vitro samples for micronuclei using their own flow cytometer. Each kit contains all of the quality-assured supplies & reagents to analyze 1,000 samples in 96 well plates or 200 samples in larger 1 * 1 ST
Catalog # MSPP1000/200
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together