This In Vitro MicroFlow kit allows users to analyze in vitro samples for micronuclei using their own flow cytometer. Each kit contains all of the quality-assured supplies & reagents to analyze 250 samples in 96 well plates or 50 samples in larger for 1 * 1 ST
Catalog # MSPP250/50
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together