Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Human OSCAR, His Tag (OSR-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Asn 229 (Accession # Q8IYS5-2). 1 * 100 µG
Catalog # ACRBOSR-H52H3100UG
Supplier:
ACRO Biosystems
Expand image
Human OSCAR, His Tag (OSR-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Asn 229 (Accession # Q8IYS5-2). 1 * 100 µG
Catalog # ACRBOSR-H52H3100UG
Supplier:
ACRO Biosystems
Frequently Bought Together
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
VWR®, Conical Sample Containers, with Snap-On Lids
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection