ContactUsLinkComponent
Biotinylated Human BTN2A1, Fc,Avitag (BT1-H52F5) is expressed from human 293 cells (HEK293). It contains AA Gln 29 - Ala 248 (Access ion # Q7KYR7-2). 1 * 200 µG
Catalog # ACRBBT1-H52F5200UG
Supplier:  ACRO Biosystems
Biotinylated Human BTN2A1, Fc,Avitag (BT1-H52F5) is expressed from human 293 cells (HEK293). It contains AA Gln 29 - Ala 248 (Access ion # Q7KYR7-2). 1 * 200 µG
Catalog # ACRBBT1-H52F5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together