ContactUsLinkComponent
Monkeypox virus (strain Zaire-96-I-16) A30L, His Tag (A3L-M5243) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Leu 146 (Accession # Q8V4U9-1). 1 * 50 µG
Catalog # ACRBA3L-M5243-50UG
Supplier:  ACROBIOSYSTEMS
Monkeypox virus (strain Zaire-96-I-16) A30L, His Tag (A3L-M5243) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Leu 146 (Accession # Q8V4U9-1). 1 * 50 µG
Catalog # ACRBA3L-M5243-50UG
Supplier:  ACROBIOSYSTEMS