ContactUsLinkComponent
Monkeypox virus (strain Zaire-96-I-16) E8L, His Tag (E8L-M52H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - T hr 275 (Accession # Q8V4Y0-1). 1 * 50 µG
Catalog # ACRBE8L-M52H3-50UG
Supplier:  ACROBIOSYSTEMS
Monkeypox virus (strain Zaire-96-I-16) E8L, His Tag (E8L-M52H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - T hr 275 (Accession # Q8V4Y0-1). 1 * 50 µG
Catalog # ACRBE8L-M52H3-50UG
Supplier:  ACROBIOSYSTEMS