ContactUsLinkComponent
Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag (BA.4/Omicron) (SPN-C82Et) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (T19I, LPP24-26del, A27S, HV69-70del, G142D, V213G, G339D, S371F, S373P, 1 * 25 µG
Catalog # ACRBSPN-C82ET-25UG
Supplier:  ACRO Biosystems
Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag (BA.4/Omicron) (SPN-C82Et) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (T19I, LPP24-26del, A27S, HV69-70del, G142D, V213G, G339D, S371F, S373P, 1 * 25 µG
Catalog # ACRBSPN-C82ET-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together