SARS-CoV-2 Spike RBD, His Tag (BA.2/Omicron) (SPD-C522n) is expressed from human 293 cells (HEK293). The spike mutations are identif ied on the SARS-CoV-2 Omicron variant (Pango lineage: BA.2 GISAID clade: GRA Nextstrain clade: 21L). 1 * 1 mg
Catalog # ACRBSPD-C522N-1MG
Supplier: ACRO Biosystems
Frequently Bought Together