ContactUsLinkComponent
SARS-CoV-2 Spike RBD, His Tag (SPD-C5228) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (Y449H, E484K, N501Y)). The mutations Y449H, E484K, N501Y were identified in the SARS-CoV-2 variant C.1.2. 1 * 100 µG
Catalog # ACRBSPD-C5228100UG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 Spike RBD, His Tag (SPD-C5228) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (Y449H, E484K, N501Y)). The mutations Y449H, E484K, N501Y were identified in the SARS-CoV-2 variant C.1.2. 1 * 100 µG
Catalog # ACRBSPD-C5228100UG
Supplier:  ACROBIOSYSTEMS