ContactUsLinkComponent
SARS-CoV-2 Spike Trimer, His Tag (SPN-C52Hb) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Acce ssion # QHD43416.1). The mutations C136F, Y144del, R190S, D215G, LA242-243del, Y449H, E484K, N501Y, D614G, H655Y, N67 1 * 50 µG
Catalog # ACRBSPN-C52HB-50UG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 Spike Trimer, His Tag (SPN-C52Hb) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Acce ssion # QHD43416.1). The mutations C136F, Y144del, R190S, D215G, LA242-243del, Y449H, E484K, N501Y, D614G, H655Y, N67 1 * 50 µG
Catalog # ACRBSPN-C52HB-50UG
Supplier:  ACROBIOSYSTEMS