ContactUsLinkComponent
Cynomolgus ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (IT8-C52W9) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Pro 993 (ITGAV) & Glu 43 - Ser 681 (ITGB8) (Accession # A0A2K5WCD3-1 (ITGAV) & G7P0S0-1 (ITGB8)). 1 * 100 µG
Catalog # ACRBIT8-C52W9100UG
Supplier:  ACROBIOSYSTEMS
Cynomolgus ITGAV&ITGB8 Heterodimer Protein, His Tag&Tag Free (IT8-C52W9) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Pro 993 (ITGAV) & Glu 43 - Ser 681 (ITGB8) (Accession # A0A2K5WCD3-1 (ITGAV) & G7P0S0-1 (ITGB8)). 1 * 100 µG
Catalog # ACRBIT8-C52W9100UG
Supplier:  ACROBIOSYSTEMS