ContactUsLinkComponent
Human IFN-alpha / beta R1 Protein, His Tag (IF1-H5225) is expressed from human 293 cells (HEK293). It contains AA Lys 28 - Lys 436 ( Accession # P17181-1). 1 * 100 µG
Catalog # ACRBIF1-H5225100UG
Supplier:  ACRO Biosystems
Human IFN-alpha / beta R1 Protein, His Tag (IF1-H5225) is expressed from human 293 cells (HEK293). It contains AA Lys 28 - Lys 436 ( Accession # P17181-1). 1 * 100 µG
Catalog # ACRBIF1-H5225100UG
Supplier:  ACRO Biosystems
Frequently Bought Together