ContactUsLinkComponent
Biotinylated Human IL-1RL1, His,Avitag™ (IL1-H82E9) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Pro 323 (Acc ession # Q01638-1). 1 * 25 µG
Catalog # ACRBIL1-H82E9-25UG
Supplier:  ACRO Biosystems
Biotinylated Human IL-1RL1, His,Avitag™ (IL1-H82E9) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - Pro 323 (Acc ession # Q01638-1). 1 * 25 µG
Catalog # ACRBIL1-H82E9-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together