ContactUsLinkComponent
HPLC COLUMN ReproSil 100 C18 5µm 15% endc. 300*3.9mm 1 * 1 items
Catalog # MAISR15.96.S3039
Supplier:  DR MAISCH
HPLC COLUMN ReproSil 100 C18 5µm 15% endc. 300*3.9mm 1 * 1 items
Catalog # MAISR15.96.S3039
Supplier:  DR MAISCH
Frequently Bought Together