Sodium Hydroxide Monohydrate (99.99%-Na) 1 * 10 g
Catalog # STRE93-1064-10G
Supplier: Strem Chemicals, Inc.
Sodium Hydroxide Monohydrate (99.99%-Na) 1 * 10 g
Catalog # STRE93-1064-10G
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Sodium hydrogen carbonate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Magnesium sulphate heptahydrate ≥98%, purified
Supplier: VWR Chemicals
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium nitrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Magnesium chloride hexahydrate 98.0-101.0% USP, Ph. Eur., BP
Supplier: VWR Chemicals