Colorclear Solution (Sodium Borohydride 1 * 500 g
Catalog # STRE11-1123-500G
Supplier: Strem Chemicals, Inc.
Colorclear Solution (Sodium Borohydride 1 * 500 g
Catalog # STRE11-1123-500G
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Sodium hydrogen carbonate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.0-100.5% USP, Ph. Eur., BP
Supplier: VWR Chemicals
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals
Potassium chloride, EMSURE® for analysis, Supelco®
Supplier: Merck
Potassium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals