Colorclear Solution (Sodium Borohydride 1 * 2,5 kg
Catalog # STRE11-1123-2500G
Supplier: Strem Chemicals, Inc.
Colorclear Solution (Sodium Borohydride 1 * 2,5 kg
Catalog # STRE11-1123-2500G
Supplier: Strem Chemicals, Inc.
Frequently Bought Together
Sodium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ammonium iron(III) sulphate dodecahydrate 99.0-101.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium chloride 99.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Ammonium acetate ≥98.0%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Sodium thiosulphate, Supelco®
Supplier: Merck
Sodium salicylate, EMSURE® for analysis, Supelco®
Supplier: Merck
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Potassium fluoride ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals