Orders LinkContactUsLinkComponent
Masterflex® Reducer, Female Threaded Adapter, Straight, Avantor® | Avantor
Masterflex® Reducer, Female Threaded Adapter, Straight, Avantor®
Fitting, 316 Stainless Steel, Straight, Female Thread Reducer, ⅛" × ½" NPT(F), Threaded Reducer, Stainless Steel 316, Straight, ½", ⅛"
Catalog # MFLX31410-13
Masterflex® Reducer, Female Threaded Adapter, Straight, Avantor®
Catalog # MFLX31410-13
Masterflex® Reducer, Female Threaded Adapter, Straight, Avantor®
Masterflex® Reducer, Female Threaded Adapter, Straight, Avantor®
Fitting, 316 Stainless Steel, Straight, Female Thread Reducer, ⅛" × ½" NPT(F), Threaded Reducer, Stainless Steel 316, Straight, ½", ⅛"
Wechseln zu:
Price
132,00 €
Anmelden für die Verfügbarkeit
1 * 1 ST
Product Details & Documents
Maintain a permanent connection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Sturdy threaded adapters are designed for permanent installation. Choose the size and material that fits your application needs.
Spezifikationen:
  • Connection Type:
    Threaded Reducer
  • Flow pattern:
    Straight
  • Thread size NPT(F) (port 1):
    ½"
  • Thread size NPT(F) (port 2):
    ⅛"
  • Description:
    Fitting, 316 Stainless Steel, Straight, Female Thread Reducer, ⅛" × ½" NPT(F)
  • Seq:
    09
  • Fitting material:
    Stainless Steel 316
  • Cat. No.:
    MFLX31410-13
  • Warranty:
    1 year
  • Model:
    31410-13
  • Sterilisation:
    Autoclave, ethylene oxide (ETO), or gamma
  • Fitting depth:
    8,73 mm