ContactUsLinkComponent
AAVS1-Puro-DNRDonor Vector for Transgene Insertion into the AAVS1 Safe Harbor Locus (Supercoil Plasmid DNA) 1 * 10 µG
Catalog # BCATGE100024-OR
Supplier:  Bio-Cat
AAVS1-Puro-DNRDonor Vector for Transgene Insertion into the AAVS1 Safe Harbor Locus (Supercoil Plasmid DNA) 1 * 10 µG
Catalog # BCATGE100024-OR
Supplier:  Bio-Cat