METHYL 4-(BROMOMETHYL)-3-(TRIFLUOROMETHYL)BENZOATE,95% 1 * 1 g
Catalog # ASTAF52483-1G
Supplier: ASTATECH
METHYL 4-(BROMOMETHYL)-3-(TRIFLUOROMETHYL)BENZOATE,95% 1 * 1 g
Catalog # ASTAF52483-1G
Supplier: ASTATECH
Frequently Bought Together
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Water analytical reagent
Supplier: Bernd Kraft
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Ethanolamine ACS
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for HPLC
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
1,10-Phenanthroline monohydrate, Supelco®
Supplier: Merck
Dimethyl sulphoxide ≥99.9% (by GC), CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Formamide 99.5+%, extra pure
Supplier: Thermo Fisher Scientific
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals