Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SkillPak GigaCap S-650M 5x1mL col., 7.0x25mm 1 * 1 items
Catalog # TOYO0045275
Supplier:
TOSOH Bioscience
Expand image
SkillPak GigaCap S-650M 5x1mL col., 7.0x25mm 1 * 1 items
Catalog # TOYO0045275
Supplier:
TOSOH Bioscience