Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Cole-Parmer Flowmeter 1-10 Gpm Water 1''Npt Acrylic,Pvc Ftg,Ss Float,150Psig.150F 1 * 1 items
Catalog # MFLX61550-00
Supplier:
Avantor Fluid Handling
Expand image
Cole-Parmer Flowmeter 1-10 Gpm Water 1''Npt Acrylic,Pvc Ftg,Ss Float,150Psig.150F 1 * 1 items
Catalog # MFLX61550-00
Supplier:
Avantor Fluid Handling