Flux Pumps Air-Powered Drum Pump Motor, 1/2 HP, 9000 rpm, on/off; 17 cfm@85 psi
Catalog # MFLX07070-70
Supplier: Avantor Fluid Handling
CE Compliant
3A Sanitary approval
About this item
Configurable to your requirements.
- High-viscosity tube set handles fluids with viscosities to 50,000 cp!
Air-operated motors are ideal for hazardous duty applications or where electricity is not available. Models 07070-60 and -70 are fitted with a muffler and include a grounding post. Model 07070-70 also has a unique trigger valve handle—press switch for intermittent pumping; lock switch for continuous use.
- Power:Air
- Max. viscosity:1200 cP
- Description:Air-Powered Drum Pump Motor, 1/2 HP, 9000 rpm, on/off; 17 cfm@85 psi (with trigger)
- Environmentally Preferable:
- Motor hp:1/2
- Warranty:6 Months
- Pk:1
- Max. motor RPM:9000 rpm