Order Entry
Germany
ContactUsLinkComponent
Flux Pumps Air-Powered Drum Pump Motor, 1/2 HP, 9000 rpm; 17 cfm@85 psi
Flux Pumps Air-Powered Drum Pump Motor, 1/2 HP, 9000 rpm; 17 cfm@85 psi
Catalog # MFLX07070-60
3A Sanitary approval
Flux Pumps Air-Powered Drum Pump Motor, 1/2 HP, 9000 rpm; 17 cfm@85 psi
Catalog # MFLX07070-60
3A Sanitary approval

About this item

Configurable to your requirements.

  • High-viscosity tube set handles fluids with viscosities to 50,000 cp!

Air-operated motors are ideal for hazardous duty applications or where electricity is not available. Models 07070-60 and -70 are fitted with a muffler and include a grounding post. Model 07070-70 also has a unique trigger valve handle—press switch for intermittent pumping; lock switch for continuous use.

  • Power:
    Air
  • Max. viscosity:
    1200 cP
  • Description:
    Air-Powered Drum Pump Motor, 1/2 HP, 9000 rpm; 17 cfm@85 psi (without trigger)
  • Environmentally Preferable:
  • Motor hp:
    1/2
  • Air consumption:
    28,9 m³/h (17 ft.³/min)
  • Warranty:
    6 Months
  • Pk:
    1
  • Max. motor RPM:
    9000 rpm