Masterflex® Tripod Base for Three to Five Tube Systems
Catalog # MFLX03226-40
Supplier: Avantor Fluid Handling
About this item
Easily mount flowmeters on tripod for easy viewing.
- Constructed of durable acrylic
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.
- Description:Tripod Base for Three to Five Tube Systems
- Seq:04
- Environmentally Preferable:
- Model:03226-40
- Width:1755"
- Warranty:1 year
- Pk:1
- Housing material:Acrylic