ContactUsLinkComponent
SPME Fiber DVB/C-WR/PDMS (Divinylbenzene/Carbon Wide Range/Polydimethylsiloxane Fiber Fiber thickness 50/30 Fiber 10mm color code: dark gray Pack of 3 1 * 3 items
Catalog # REST27873.3
Supplier:  Restek
SPME Fiber DVB/C-WR/PDMS (Divinylbenzene/Carbon Wide Range/Polydimethylsiloxane Fiber Fiber thickness 50/30 Fiber 10mm color code: dark gray Pack of 3 1 * 3 items
Catalog # REST27873.3
Supplier:  Restek