Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Class VI PS Coupling Insert Cap with Class VI Silicone O- Ring and Lock - Bag of 25 1 * 25 items
Catalog # CPCCMPXK32039B25
Supplier:
CPC COLDER
Expand image
Class VI PS Coupling Insert Cap with Class VI Silicone O- Ring and Lock - Bag of 25 1 * 25 items
Catalog # CPCCMPXK32039B25
Supplier:
CPC COLDER