Grounding cables, b.safe
Grounding cable with ring cable plug M6 and 32 mm Ø tube clip, 3,0 m
Catalog # BOHLR607-03
Supplier: Bohlender
Jump to:
Product Details & Documents
For earth connection of vessels and components made of static dissipative materials.
- Choose the suitable connection to earth that we offer in our range of grounding cables with different connectors
- Customised versions can be offered on request
- Suitable grounding accessories for b.safe products
The scope of delivery of the ring cable lug includes screw, nut and washer.
Specifications
- Description:Grounding cable with ring cable plug M6 and 32 mm Ø tube clip
- Length:3,0 m
- Seq:11



