ContactUsLinkComponent
SARS-CoV-2 Spike Trimer His Tag (B.1.1.529/Omicron) (SPN-C52Hz) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (A67V HV69-70del T95I G142D VYY143-145del N211del L212I ins214EPE G339D S375F Y 1 * 50 µG
Catalog # ACRBSPN-C52HZ-50UG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 Spike Trimer His Tag (B.1.1.529/Omicron) (SPN-C52Hz) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (A67V HV69-70del T95I G142D VYY143-145del N211del L212I ins214EPE G339D S375F Y 1 * 50 µG
Catalog # ACRBSPN-C52HZ-50UG
Supplier:  ACROBIOSYSTEMS