Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Trimethylamine Standard 125 ml, 1 000 µg/ml, Matrix: HCl 1 * 125 mL
Catalog # INORICTMA1-125
Supplier:
INORGANIC VENTURESIV LABS
Expand image
Trimethylamine Standard 125 ml, 1 000 µg/ml, Matrix: HCl 1 * 125 mL
Catalog # INORICTMA1-125
Supplier:
INORGANIC VENTURESIV LABS