Orders LinkContactUsLinkComponent
Volumetric flasks, class B
Volumetric flasks, class B
500 ml, 19/26, 0,50 ± ml
Catalog # VITL675941
Supplier:   VITLAB
Volumetric flasks, class B
Catalog # VITL675941
Supplier:  VITLAB
Volumetric flasks, class B
Volumetric flasks, class B
500 ml, 19/26, 0,50 ± ml
Price
33,60 €
Log in for availability
1 * 2 ST
Product Details & Documents
PP, with PP stoppers.
Highly transparent in accordance with DIN 12664 (glass) with blue ring mark and lettering. Individually adjusted mark to contain (TC, In).
Specifications
  • NS:
    19/26
  • Capacity:
    500 ml
  • Seq:
    06
  • Tolerance:
    0,50 ± ml