Order Entry
Germany
Orders LinkContactUsLinkComponent
VWR®, Inserts for Short Thread Vials ND9

VWR®, Inserts for Short Thread Vials ND9

Micro-insert with filling lines and attached plastic spring, 0,10 ml, Conical bottom, 6×29 mm, PP, Transparent, No
Catalog # 548-1381A
Supplier:   VWR Collection
VWR®, Inserts for Short Thread Vials ND9
Catalog # 548-1381A
Supplier:  VWR Collection
VWR®, Inserts for Short Thread Vials ND9
VWR®, Inserts for Short Thread Vials ND9

Micro-insert with filling lines and attached plastic spring, 0,10 ml, Conical bottom, 6×29 mm, PP, Transparent, No

Price
750,00 €
Log in for availability
1 * 1.000 ST
Specifications
  • Cap included:
    No
  • Ø×H:
    6×29 mm
  • Capacity:
    0,10 ml
  • Colour:
    Transparent
  • Description:
    Micro-insert with filling lines and attached plastic spring
  • Material:
    PP
  • Seq:
    06
  • Base:
    Conical bottom
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.