Order Entry
Germany
Orders LinkContactUsLinkComponent
VWR®, Inserts for Short Thread Vials ND9

VWR®, Inserts for Short Thread Vials ND9

Micro-insert with assembled plastic spring, 0,10 ml, Conical bottom, 5,7×29 mm, Borosilicate glass 5.1, Clear, No
Catalog # 548-0002A
Supplier:   VWR Collection
VWR®, Inserts for Short Thread Vials ND9
Catalog # 548-0002A
Supplier:  VWR Collection
VWR®, Inserts for Short Thread Vials ND9
VWR®, Inserts for Short Thread Vials ND9

Micro-insert with assembled plastic spring, 0,10 ml, Conical bottom, 5,7×29 mm, Borosilicate glass 5.1, Clear, No

Price
1 420,00 €
Log in for availability
1 * 1.000 ST
Specifications
  • Cap included:
    No
  • Ø×H:
    5,7×29 mm
  • Capacity:
    0,10 ml
  • Colour:
    Clear
  • Description:
    Micro-insert with assembled plastic spring
  • Material:
    Borosilicate glass 5.1
  • Seq:
    02
  • Base:
    Conical bottom
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.