Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
The flow rate can be calibrated between 0,25 to 5 mL/min. Flow-Splitter Split Ratio 1:1 to 20:1 maximum pressure: 350 bar / 5000 psi 1 * 1 ST
Catalog # MSPPASI610-PO10-06
Supplier:
MARKET SOURCE PART PROCESS
Expand image
The flow rate can be calibrated between 0,25 to 5 mL/min. Flow-Splitter Split Ratio 1:1 to 20:1 maximum pressure: 350 bar / 5000 psi 1 * 1 ST
Catalog # MSPPASI610-PO10-06
Supplier:
MARKET SOURCE PART PROCESS